Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Peptide Sequence: MAGMPGTAET FETPETCGTT DMRDSRDPMY RREGSYDRYL RMDDYCRRKD DSYFDRYRDS FDGRGPPGPE SQSRAKERLK REERRREELY RQYFEEIQRR FDAERPVDCS VIVVNKQTKD YAESVGRKVR DLGMVVDLIF LNTEVSLSQA LEDVSRGGSP FAIVITQQHQ IHRSCTVNIM FGTPQEHRNM PQADAMVLVA RNYERYKNEC REKEREEIAR QAAKMADEAI LQERERGGPE EGVRGGHPPA IQSLINLLAD NRYLTAEETD KIINYLRERK ERLMRSSTDS LPGELRGRAE ARFPANHSGR PRVPR
This gene encodes a coregulator for the alpha and beta estrogen receptors and the orphan nuclear receptor NR1D2. The protein localizes to the nucleus, and is thought to have both coactivator and corepressor functions. Its interaction with nuclear receptors is independent of the AF2 domain on the receptors, which is known to regulate interaction with other coreceptors. Several alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coactivator independent of activation function 2; coactivator independent of AF-2; NCoA-5
Gene Aliases: bA465L10.6; CIA
UniProt ID: (Human) Q9HCD5
Entrez Gene ID: (Human) 57727
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support