Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
GTPase-activating protein for ARF1 and, to a lesser extent, ARF5. Directly and specifically regulates the adapter protein 3 (AP-3)-dependent trafficking of proteins in the endosomal-lysosomal system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGAP-1; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 1; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 1; centaurin, gamma 2; Centaurin-gamma-2; Cnt-g2; GGAP1; GTP-binding and GTPase-activating protein 1; unnamed protein product
Gene Aliases: AGAP-1; AGAP1; CENTG2; cnt-g2; GGAP1; KIAA1099
UniProt ID: (Human) Q9UPQ3
Entrez Gene ID: (Human) 116987
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support