Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Keep as concentrated solution.
Predicted reactivity: Mouse (98%), Rat (98%), Chicken (97%), Bovine (99%).
Positive Control: 293T, A431, HeLa, HepG2.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FOXK1 is expressed in these cells and regulates cell cycle progression through an interaction with its downstream target the cyclin-dependent kinase inhibitor p21 (CIP). Loss of FOXK1 in mice results in growth retardation and a severe impairment in skeletal muscle regeneration following injury. FOXK1 also shows expression in immature tissues of brain, eye, heart, lung and thymus. It also is predominantly expressed in many malignant tissues, such as tumors of the brain, colon and lymph node.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABP-280 homolog; actin binding protein 278; actin-binding-like protein; beta-filamin; filamin B, beta; filamin homolog 1; filamin, beta; filamin-3; filamin-B-like; FilaminB; homolog of ABP-280; Larsen syndrome 1 (autosomal dominant); thyroid autoantigen
Gene Aliases: ABP-278; ABP-280; FH1; FLN-B; FLN1L; LRS1; TABP; TAP
UniProt ID: (Human) O75369
Entrez Gene ID: (Human) 2317, (Rat) 306204
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support