Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAAATGDPGL SKLQFAPFSS ALDVGFWHEL TQKKLNEYRL DEAPKDIKGY YYNGDSAGLP ARLTLEFSAF DMSAPTPARC CPAIGTLYNT NTLESFKTAD
This gene was identified based on homology to Pichia pastoris GSA7 and Saccharomyces cerevisiae APG7. In the yeast, the protein appears to be required for fusion of peroxisomal and vacuolar membranes. The protein shows homology to the ATP-binding and catalytic sites of the E1 ubiquitin activating enzymes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APG7; APG7 autophagy 7-like; AT5G45900; ATAPG7; ATATG7; ATG12-activating enzyme E1 ATG7; ATG7 autophagy related 7 homolog; AUTOPHAGY 7; autophagy 7-like; autophagy related 7 homolog; AUTOPHAGY-RELATED 7; autophagy-related protein 7; DKFZp434N0735; hAGP7; K15I22.10; K15I22_10; PEROXISOME UNUSUAL POSITIONING 4; PEUP4; ubiquitin-activating enzyme E1-like protein; unnamed protein product
Gene Aliases: 1810013K23Rik; Agp7; APG7-LIKE; APG7L; Atg7l; Gm21553; GSA7; SCAR31
UniProt ID: (Human) O95352, (Mouse) Q9D906, (Rat) Q641Y5
Entrez Gene ID: (Human) 10533, (Mouse) 74244, (Rat) 312647
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support