Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Positive Samples: Mouse liver, Rat heart
Immunogen sequence: VFGEIEECTV NLRDDGDSYG FITYRYTCDA FAALENGYTL RRSNETDFEL YFCGRKQFFK SNYADLDSNS DDFDPASTKS KYDSLDFDSL LKEAQRSLRR
PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERR ligand 1; L-PGC-1alpha; ligand effect modulator-6; PERC; PERC-s; lacks one estrogen receptor interaction domain; peroxisome proliferative activated receptor gamma coactivator 1; peroxisome proliferative activated receptor, gamma, coactivator 1 alpha; peroxisome proliferative activated receptor, gamma, coactivator 1 beta; peroxisome proliferator-activated receptor gamma coactivator 1beta-2a; peroxisome proliferator-activated receptor gamma, coactivator 1 beta; PGC-1; pgc-1 alpha; pgc-1 beta; PGC-1-alpha; PGC-1-beta; PGC-1-related estrogen receptor alpha coactivator; PGC-1alpha; PGC1beta-1a; PGC1beta-1b; PGC1beta-2a; PGC1beta-2b; PPAR gamma coactivator 1alpha variant form-1; PPAR gamma coactivator variant form; PPAR gamma coactivator-1beta protein; PPAR-gamma coactivator 1-alpha; PPAR-gamma coactivator 1-beta; PPARAGCIalpha; PPARAGCIbeta; PPARgamma coactivator 1 beta; PPARgamma coactivator 1alpha; PPARGC-1-alpha; PPARGC-1-beta; Ppargc1a-vf1; transcriptional coactivator; transcriptional coactivator; PGC-1; unnamed protein product
Gene Aliases: 4631412G21Rik; A830037N07Rik; ERRL1; Gm11133; LEM6; LRPGC1; PERC; PGC-1; PGC-1(alpha); PGC-1(beta); PGC-1alpha; PGC-1beta; PGC-1beta/ERRL1; PGC-1v; PGC1; PGC1A; PGC1B; PGC1beta; Pgco1; PGCvf; PGCvf-1; PGCvf1; PPARGC-1-alpha; PPARGC-1-beta; PPARGC1
UniProt ID: (Human) Q9UBK2, (Human) Q86YN6, (Mouse) O70343, (Rat) Q9QYK2, (Mouse) Q8VHJ7, (Rat) Q811R2
Entrez Gene ID: (Human) 10891, (Human) 133522, (Mouse) 19017, (Rat) 83516, (Mouse) 170826, (Rat) 291567
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support