Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • PCAF Antibodies

          Invitrogen

          PCAF Monoclonal Antibody (7D12C8)

          View all (31) PCAF antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PCAF Monoclonal Antibody (7D12C8)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          PCAF Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PCAF Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PCAF Antibody (MA5-38547) in ICC/IF

          Immunocytochemistry/Immunofluorescence analysis of PCAF in HeLa cells using PCAF Monoclonal Antibody (Product # MA5-38547) (green). Blue: DRAQ5 fluorescent DNA dye. Red: Actin filaments have been labeled with Alexa Fluor- 555 phalloidin. Secondary antibody from Fisher (Product # 35503) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PCAF Antibody in Immunocytochemistry (ICC/IF)
          PCAF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PCAF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PCAF Antibody in Western Blot (WB)
          PCAF Antibody in Western Blot (WB)
          PCAF Antibody in Flow Cytometry (Flow)
          PCAF Monoclonal Antibody (7D12C8)

          Product Details

          MA5-38547

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:200-1:1,000
          -

          Flow Cytometry (Flow)

          1:200-1:400
          -

          ELISA (ELISA)

          1:10,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          7D12C8

          Immunogen

          Purified recombinant fragment of human KAT2B (aa: 1-142) expressed in E. Coli.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2898459

          Product Specific Information

          This antibody has been tested in indirect ELISA.

          Target Information

          PCAF (histone acetyltransferase KAT2B) functions as a histone acetyltransferase (HAT) to promote transcriptional activation. It has significant histone acetyltransferase activity with core histones (H3 and H4) and nucleosome core particles. PCAF also acetylates non-histone proteins such as ACLY, PLK4, and TBX5. It inhibits cell-cycle progression and counteracts the mitogenic activity of the adenoviral oncoprotein E1A. PCAF also acts as a circadian transcriptional coactivator which enhances the activity of the circadian transcriptional activators NPAS2-ARNTL/BMAL1 and CLOCK-ARNTL/BMAL1 heterodimers. It is involved in heart and limb development by mediating acetylation of TBX5 and acetylation regulating nucleocytoplasmic shutting of TBX5.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CREBBP-associated factor; Histone acetylase PCAF; Histone acetyltransferase KAT2B; Histone acetyltransferase PCAF; K(lysine) acetyltransferase 2B; Lysine acetyltransferase 2B; P/ PCAF; P/CAF; P300/CBP-associated factor; Spermidine acetyltransferase KAT2B

          View more View less

          Gene Aliases: A930006P13Rik; AI461839; AW536563; CAF; KAT2B; P/CAF; PCAF

          View more View less

          UniProt ID: (Human) Q92831, (Mouse) Q9JHD1

          View more View less

          Entrez Gene ID: (Human) 8850, (Rat) 301164, (Mouse) 18519

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding chromatin binding transcription cofactor activity transcription coactivator activity diamine N-acetyltransferase activity histone acetyltransferase activity lysine N-acetyltransferase activity, acting on acetyl phosphate as donor cyclin-dependent protein serine/threonine kinase inhibitor activity protein binding enzyme activator activity H3 histone acetyltransferase activity acetyltransferase activity transferase activity transferase activity, transferring acyl groups transferase activity, transferring acyl groups other than amino-acyl groups protein kinase binding histone acetyltransferase binding histone deacetylase binding histone acetyltransferase activity (H3-K9 specific) peptide-lysine-N-acetyltransferase activity DNA-binding transcription factor binding transcription factor binding protein complex binding N-acetyltransferase activity
          Process(es)
          chromatin remodeling protein acetylation memory negative regulation of cell proliferation regulation of protein ADP-ribosylation positive regulation of neuron projection development histone acetylation N-terminal peptidyl-lysine acetylation internal peptidyl-lysine acetylation peptidyl-lysine acetylation cellular response to insulin stimulus cellular response to oxidative stress positive regulation of chromatin binding positive regulation of gluconeogenesis by positive regulation of transcription from RNA polymerase II promoter histone H3 acetylation histone H3-K9 acetylation negative regulation of cyclin-dependent protein serine/threonine kinase activity positive regulation of transcription, DNA-templated positive regulation of vasodilation positive regulation of transcription from RNA polymerase II promoter cellular response to parathyroid hormone stimulus positive regulation of histone H3-K14 acetylation positive regulation of histone H3-K9 acetylation negative regulation of transcription from RNA polymerase II promoter positive regulation of transcription from RNA polymerase II promoter by glucose gluconeogenesis regulation of DNA repair regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter ubiquitin-dependent protein catabolic process fatty acid biosynthetic process heart development regulation of gene expression vasodilation regulation of RNA splicing negative regulation of fatty acid biosynthetic process positive regulation of gluconeogenesis positive regulation of fatty acid biosynthetic process positive regulation of gene expression, epigenetic positive regulation of glycolytic process regulation of embryonic development negative regulation of centriole replication positive regulation of lipid biosynthetic process rhythmic process regulation of cell division attachment of mitotic spindle microtubules to kinetochore regulation of cell cycle limb development regulation of small molecule metabolic process negative regulation of ferroptosis positive regulation of attachment of mitotic spindle microtubules to kinetochore negative regulation of rRNA processing transcription, DNA-templated cell cycle
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0