Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
C1-inhibitor (C1INH) functions as a regulator of the activation of the classical complement pathway and of the contact activation system of kinin generation and coagulation. Its primary biologically relevant target proteinases are C1r, C1s, coagulation factors XIa and XIIa, and plasma kallikrein. C1-inhibitor consists of one polypeptide chain and has a molecular mass of ~104 kDa. Serum concentration has been determined to 180 µg/mL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1-inactivator; C1-inhibiting factor; C1-inhibitor; C1Inh; complement component 1 inhibitor; epididymis secretory sperm binding protein; esterase inhibitor; IC1; plasma protease (C1) inhibitor precursor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine/cysteine proteinase inhibitor clade G member 1; serpin G1; serpin peptidase inhibitor clade G (C1 inhibitor) member 1 isoform SERPING1_1; serpin peptidase inhibitor clade G (C1 inhibitor) member 1 isoform SERPING1_2; serpin peptidase inhibitor clade G (C1 inhibitor) member 1 isoform SERPING1_3; serpin peptidase inhibitor clade G (C1 inhibitor) member 1 isoform SERPING1_4; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serpin peptidase inhibitor, clade G, member 1; SERPING1; unnamed protein product
Gene Aliases: C1-IA; C1-INH; C1IN; C1INH; C1NH; HAE1; HAE2
UniProt ID: (Human) P05155, (Rat) Q6P734
Entrez Gene ID: (Pig) 100144304, (Human) 710, (Rat) 295703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support