Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Positive test controls include: HeLa, 293T, NIH/3T3, K-562, Mouse testis, Mouse heart, Rat testis. The target is usually found in the following locations: Cell membrane, Nucleus.
Immunogen sequence: FKLVIEFSEE YPNKPPTVRF LSKMFHPNVY ADGSICLDIL QNRWSPTYDV SSILTSIQSL LDEPNPNSPA NSQAAQLYQE NKREYEKRVS AIVEQSWNDS
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2B is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Its protein sequence is 100% identical to the mouse, rat, and rabbit homologs, which indicates that this enzyme is highly conserved in eukaryotic evolution.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E2 protein; E2 ubiquitin-conjugating enzyme B; E2(14k); HHR6B (Human homologue of yeast RAD 6); putative; RAD6 homolog B; UBE2B; ubiquitin carrier protein B; ubiquitin conjugating enzyme; ubiquitin conjugating enzyme E2B; ubiquitin-conjugating enzyme E2(14k); ubiquitin-conjugating enzyme E2-17 kDa; ubiquitin-conjugating enzyme E2B (RAD6 homolog); ubiquitin-conjugating enzyme E2B (RAD6 homolog, S. cerevisiae); ubiquitin-conjugating enzyme E2B, RAD6 homolog; ubiquitin-conjugating enzyme E2B, RAD6 homology; ubiquitin-protein ligase B; unnamed protein product
Gene Aliases: E2-14k; E2-17kDa; E214K; HHR6B; HR6B; mHR6B; RAD6B; UBC2
UniProt ID: (Human) P63146, (Mouse) P63147, (Rat) P63149
Entrez Gene ID: (Human) 7320, (Mouse) 22210, (Rat) 81816
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support