Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • HSP70/HSC70 Antibodies

          Invitrogen

          HSP70/HSC70 Monoclonal Antibody (N27F3-4), PerCP

          View all (8) HSP70/HSC70 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite HSP70/HSC70 Monoclonal Antibody (N27F3-4), PerCP

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          HSP70/HSC70 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          HSP70/HSC70 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HSP70/HSC70 Antibody (MA5-45095) in IHC (P)

          Immunohistochemistry analysis using backskin. Fixation involved Bouins Fixative and paraffin-embedded. Samples were incubated with Hsp70 monoclonal antibody (Product # MA5-45095) at 1:100 for 1 hour at RT, followed by FITC Goat Anti-Mouse (green) at 1:50 for 1 hour at RT. Localization: Epidermis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HSP70/HSC70 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSP70/HSC70 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA5-45095

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100
          -

          Immunocytochemistry (ICC/IF)

          1:50
          -

          Flow Cytometry (Flow)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          Immunomicroscopy (IM)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, C. elegans, Chicken, Fruit fly, Fish, Guinea pig, Hamster, Human, Mammal, Mouse, Non-human primate, Sheep, Plant, Pig, Rabbit, Rat, Xenopus

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N27F3-4

          Immunogen

          Recombinant HSP70/HSC70

          Conjugate

          PerCP PerCP PerCP
          • APC 
          • FITC
          • Request custom conjugation

          Excitation/Emission Max

          482/675 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          200 µg

          Purification

          Protein G

          Storage buffer

          95.64mM phosphate/2.48mM MES, pH 7.2, with 0.5M EDTA

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2931549

          Product Specific Information

          1 µg/mL of MA5-45095 was sufficient for detection of HSP70/HSC70 in 20 µg of heat shocked HeLa cell lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 72 (HSP) and approximately 73kDa (HSC).

          Target Information

          HSPA4 (Heat Shock Protein Family A (Hsp70) Member 4) is a Protein Coding gene. Diseases associated with HSPA4 include Chagas Disease and Babesiosis. Among its related pathways are Regulation of activated PAK-2p34 by proteasome mediated degradation and Cellular responses to stimuli. An important paralog of this gene is HSPH1. HSPA8 (Heat Shock Protein Family A (Hsp70) Member 8) is a Protein Coding gene. Diseases associated with HSPA8 include Auditory System Disease and Borna Disease. Among its related pathways are Processing of Capped Intron-Containing Pre-mRNA and Selective autophagy. Gene Ontology (GO) annotations related to this gene include RNA binding and ubiquitin protein ligase binding. An important paralog of this gene is HSPA2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 70 kDa heat-shock-like protein; constitutive heat shock protein 70; epididymis luminal protein 33; epididymis secretory sperm binding protein Li 5a; epididymis secretory sperm binding protein Li 72p; Heat shock 70 kDa protein 4; Heat shock 70 kDa protein 8; heat shock 70-related protein APG-2; heat shock 70kd protein 10; heat shock 70kd protein 10 (HSC71); heat shock 70kD protein 4; heat shock 70kD protein 8; heat shock 70kDa protein 1-like; heat shock 70kDa protein 4; heat shock 70kDa protein 8; heat shock cognate 70; heat shock cognate 71 kD protein; heat shock cognate hsc73; heat shock cognate protein 54; Heat shock cognate protein 70; heat shock protein 4; heat shock protein 70 cognate; heat shock protein 8; heat shock protein A8; heat shock protein Apg-2; heat shock protein cognate 70; heat shock protein family A member 4; heat shock protein family A member 8; heat shock protein family H member 2; heat shock protein, 110 kDa; Hsc70 ATPase; hsp70 RY; HSPA4/HSPA8; IRP94; ischemia responsive 94 kDa protein; lipopolysaccharide-associated protein 1; LPS-associated protein 1; N-myristoyltransferase inhibitor protein 71; unnamed protein product

          View more View less

          Gene Aliases: 2410008N15Rik; 70kDa; APG-2; HEL-33; HEL-S-5a; HEL-S-72p; HS24/P52; HSC54; HSC70; hsc70.I; HSC71; Hsc73; Hsp110; HSP70; Hsp70RY; HSP71; HSP73; HSPA1; HSPA10; hspa1l; HSPH2; I79_004332; I79_022974; irp94; LAP-1; LAP1; mKIAA4025; NIP71; RY; XELAEV_18037286mg; xhsp70

          View more View less

          UniProt ID: (Dog) Q7YQC6, (Dog) Q2TFN9, (Human) P34932, (Human) P11142, (Chicken) O73885, (Bovine) P19120, (Rat) O88600, (Mouse) P63017, (Rat) P63018, (Chinese hamster) P19378

          View more View less

          Entrez Gene ID: (Dog) 403612, (Chicken) 416339, (Dog) 474680, (Sheep) 101122782, (Pig) 100524853, (Bovine) 536558, (Human) 3308, (Rabbit) 100338049, (Dog) 479406, (Sheep) 101112844, (Human) 3312, (Chicken) 395853, (Bovine) 281831, (Rabbit) 100348873, (Pig) 100511890, (Rat) 266759, (Mouse) 15525, (Guinea pig) 100735343, (Chinese hamster) 100764628, (Mouse) 15481, (Guinea pig) 100713849, (Rat) 24468, (Chinese hamster) 100689472, (Western clawed frog) 100037899, (Xenopus) 379760

          View more View less

          Function(s)
          protein binding ATP binding adenyl-nucleotide exchange factor activity ATPase activity G-protein coupled receptor binding RNA binding enzyme binding MHC class II protein complex binding protein binding, bridging heat shock protein binding ubiquitin protein ligase binding protein binding involved in protein folding cadherin binding receptor agonist activity chaperone binding C3HC4-type RING finger domain binding ATP-dependent protein disaggregase activity ATP-dependent protein folding chaperone unfolded protein binding misfolded protein binding clathrin-uncoating ATPase activity macromolecular complex binding RNA polymerase II sequence-specific DNA binding transcription factor binding phosphatidylserine binding receptor binding lipid binding A1 adenosine receptor binding AP-3 adaptor complex binding peptide binding lncRNA binding prostaglandin binding
          Process(es)
          kidney development negative regulation of protein phosphorylation protein folding negative regulation of gene expression protein import into mitochondrial matrix negative regulation of apoptotic process positive regulation of angiogenesis chaperone-mediated protein complex assembly neuron apoptotic process regulation of microglial cell activation G1/S transition of mitotic cell cycle mRNA splicing, via spliceosome positive regulation of T cell mediated cytotoxicity protein import into nucleus autophagy signal transduction skeletal muscle tissue development axo-dendritic transport response to xenobiotic stimulus response to nickel cation positive regulation of gene expression negative regulation of cardiac muscle cell apoptotic process response to activity cerebellum development protein catabolic process positive regulation of cell migration forebrain development regulation of protein stability response to estradiol response to progesterone protein-containing complex disassembly cellular response to heat protein refolding response to starvation positive regulation of catalytic activity intracellular protein transmembrane import modulation by host of viral process positive regulation by host of viral genome replication estrous cycle response to ethanol positive regulation of proteolysis negative regulation of transcription, DNA-templated ATP metabolic process protein autophosphorylation positive regulation of mRNA splicing, via spliceosome positive regulation of phagocytosis regulation of cell cycle regulation of protein complex stability chaperone-mediated autophagy late endosomal microautophagy protein targeting to lysosome involved in chaperone-mediated autophagy cellular response to hydrogen peroxide cellular response to cadmium ion clathrin coat disassembly positive regulation of lysosomal membrane permeability maintenance of postsynaptic specialization structure regulation of postsynapse organization positive regulation of ferroptosis negative regulation of NLRP3 inflammasome complex assembly negative regulation of fibril organization positive regulation of protein refolding chaperone-mediated autophagy translocation complex disassembly slow axonal transport response to odorant response to stress response to unfolded protein cellular response to starvation negative regulation of signal transduction regulation of cytoplasmic pattern recognition receptor signaling pathway regulation of programmed cell death regulation of protein complex assembly negative regulation of cellular component organization membrane organization cellular response to steroid hormone stimulus regulation of fibril organization regulation of protein import gastrulation embryo development ending in birth or egg hatching vesicle-mediated transport cellular response to unfolded protein embryonic organ development chaperone mediated protein folding requiring cofactor chaperone-mediated protein transport involved in chaperone-mediated autophagy mRNA processing RNA splicing synaptic vesicle uncoating positive regulation of microtubule nucleation regulation of mitotic spindle assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-pj9bv:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0