Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and bovine orthologs. This gene contains a 7 bp repeat sequence in its 5' UTR, and changes in the number of this repeat has been associated with early-onset schizophrenia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 14-3-3 eta; 14-3-3 protein eta chain; 14-3-3 protein eta-subtype; 14-3-3n; 1433F; AS1; brain-specific tyrosine and tryptophan hydroxylase activator; KCIP-1; protein kinase C inhibitor protein 1; similar to Homo sapiens DNA for 14-3-3 protein eta chain, exon 2 with GenBank Accession Number D78577.1; tyrosine 3-monooxygenase/tryotophan 5-monooxygenase activation protein; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide; unnamed protein product; YWHA114-3-3 eta; YWHAH
Gene Aliases: 14-3-3e; YWHA1
UniProt ID: (Human) Q04917, (Bovine) P68509, (Mouse) P68510, (Rat) P68511
Entrez Gene ID: (Dog) 100687866, (Human) 7533, (Bovine) 282126, (Mouse) 22629, (Rat) 25576
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support