Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
The Aurora gene, a serine-threonine kinase, orginally discovered in Drosophila, has been found to be responsible for a chromosomal segregation defect. The mammalian homologues of Drosophila Aurora have been termed ARK1 (Aurora-related kinase 1) and ARK2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AIM-1; ARK-2; aurora 1; aurora 2; aurora A; aurora B; aurora family kinase 1; aurora kinase B-Sv1; aurora kinase B-Sv2; aurora- and IPL1-like midbody-associated protein 1; aurora- and Ipl1-like midbody-associated protein 1 homolog; aurora-1; aurora-A; aurora-B; aurora-related kinase 1; aurora-related kinase 2; aurora/IPL1-like kinase; aurora/IPL1-related kinase 1; aurora/IPL1-related kinase 2; breast tumor-amplified kinase; IAK1; ipl1- and aurora-related kinase 1; protein kinase; protein phosphatase 1, regulatory subunit 47; protein phosphatase 1, regulatory subunit 48; ratAurA; serine/threonine kinase 12; serine/threonine kinase 5; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine-protein kinase 12; serine/threonine-protein kinase 15; serine/threonine-protein kinase 5; serine/threonine-protein kinase 6; serine/threonine-protein kinase aurora-A; serine/threonine-protein kinase aurora-B; serine/threonine-protein kinase Ayk1; STK6; aurora-A; IPL1-related kinase; AIK; BTAK; STK15; unnamed protein product
Gene Aliases: AIK; AIK2; AIM-1; AIM1; AIRK1; AIRK2; ARK-1; ARK-2; ARK1; ARK2; AURA; AurB; aurkb-sv1; aurkb-sv2; Aurora-A; Ayk1; BTAK; IAK; IAK1; IPL1; PPP1R47; PPP1R48; STK-1; STK12; STK15; STK5; STK6; STK7
UniProt ID: (Human) O14965, (Human) Q96GD4, (Mouse) P97477, (Rat) O55099, (Mouse) O70126
Entrez Gene ID: (Human) 6790, (Human) 9212, (Rat) 261730, (Mouse) 20878, (Rat) 114592, (Mouse) 20877
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support