Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Aldolases are important glycolytic cytosolic enzymes which catalyse the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate. There are three aldolase isozymes coded by three distinct genes in mammals, namely aldolases A, B, and C. Aldolase A is heavily expressed in muscle enzyme and aldolase B is heavily expressed in liver and is regarded as a liver specific enzyme. In the adult, aldolase C is the brain-specific isozyme expressed in astrocytes and a few classes of neurons, notably Purkinje cells. Appropriate antibodies to aldolase C are therefore useful to identify astrocytes in cell culture and sectioned material, and the enzyme may be over expressed in some forms of cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ALDA; ALDB; ALDC; ALDOA; ALDOC; aldolase 2; aldolase 3; aldolase A; aldolase A (EC 4.1.2.13); aldolase A (EC 4.1.3.13); aldolase A, fructose-bisphosphate; aldolase B; Aldolase B, fructose-biphosphate; aldolase B, fructose-bisphosphatase; aldolase B, fructose-bisphosphate; aldolase C, fructose-biphosphate; aldolase C, fructose-bisphosphate; brain-type aldolase; epididymis secretory sperm binding protein; epididymis secretory sperm binding protein Li 87p; fructoaldolase C; fructose 1,6-diphosphate aldolase A (EC 4.1.2.13); fructose-1,6-biphosphate triosephosphate lyase; fructose-1,6-bisphosphate triosephosphate-lyase; gluconeogenic enzyme; ALDOA_V1; gluconeogenic enzyme; ALDOA_V2; liver-type aldolase; lung cancer antigen NY-LU-1; muscle-type aldolase; scrapie responsive gene 2; scrapie-responsive protein 2; unnamed protein product; zebrin II
Gene Aliases: ALDA; ALDB; ALDC; ALDCAA; Aldo-1; Aldo-2; Aldo1; ALDO2; Aldo3; F16dip7; GSD12; HEL-S-87p; LIV10; RATALDCAA; RNALDOG5; Scrg2
UniProt ID: (Human) P04075, (Human) P05062, (Human) P09972, (Rat) P05065, (Mouse) P05064, (Mouse) Q91Y97, (Rat) P09117, (Mouse) P05063
Entrez Gene ID: (Human) 226, (Human) 229, (Human) 230, (Rat) 24189, (Mouse) 11674, (Rat) 24190, (Mouse) 230163, (Rat) 24191, (Mouse) 11676
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support