Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
The exact function of C3orf64 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AER61 glycosyltransferase; EGF domain-specific O-linked N-acetylglucosamine (GlcNAc) transferase; EGF-O-GlcNAc transferase; extracellular O-linked N-acetylglucosamine transferase; glycosyltransferase Aer61; unnamed protein product
Gene Aliases: A130022J15Rik; AER61; AOS4; C3orf64; EOGT1
UniProt ID: (Human) Q5NDL2, (Rat) Q5NDL0, (Mouse) Q8BYW9
Entrez Gene ID: (Human) 285203, (Rat) 494219, (Mouse) 101351
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support