Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Sequence of this protein is as follows: PGIIGRIGDT KSEIKILSIG GGAGEIDLQI LSKVQAQYPG VCINNEVVEP SAEQIAKYKE LVAKTSNLEN VKFAWHKETS SEYQSRMLEK KELQKWDFIH MIQMLYYVKD IPATLKFFHS LLGTNAKMLI IVVSGSSGWD KLWKKYGSRF PQDDLCQYIT SDDLTQMLDN LGLKYECYDL LSTMDISDCF IDGNENGDLL WDFLTETCNF NATAPPDLRA ELGKDLQEPE FSAKKEGKVL FNNTLSFIVI
In mammals, histamine is metabolized by two major pathways: N(tau)-methylation via histamine N-methyltransferase and oxidative deamination via diamine oxidase. HNMT is the first enzyme which is found in the cytosol and uses S-adenosyl-L-methionine as the methyl donor. In the mammalian brain, the neurotransmitter activity of histamine is controlled by N(tau)-methylation as diamine oxidase is not found in the central nervous system. A common genetic polymorphism affects the activity levels of this gene product in red blood cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HNMT-1; HNMT-2; HNMT-3; HNMT-S1; HNMT-S2; unnamed protein product
Gene Aliases: HMT; HNMT-S1; HNMT-S2; MRT51
UniProt ID: (Human) P50135
Entrez Gene ID: (Human) 3176
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support