Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 15-hydroxyprostaglandin dehydrogenase; 20-beta-hydroxysteroid dehydrogenase; alcohol dehydrogenase [NAD(P)+] CBR1; carbonyl reductase (NADPH) 1; CBR1; epididymis secretory sperm binding protein; NADPH-dependent carbonyl reductase 1; prostaglandin 9-ketoreductase; prostaglandin-E(2) 9-reductase; short chain dehydrogenase/reductase family 21C member 1; unnamed protein product
Gene Aliases: CBR; hCBR1; PG-9-KR; SDR21C1
UniProt ID: (Human) P16152, (Rat) P47727
Entrez Gene ID: (Human) 873, (Rat) 29224
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support