Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
DPYSL3 is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. DPYSL3 plays a role in axon guidance, neuronal growth cone collapse and cell migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 75 kDa isoform of TUC-4; collapsin response mediator protein 4; collapsin response mediator protein 4 long; CRMP-4; CRMP4A; DRP-3; LCRMP-4; testicular secretory protein Li 7; TOAD-64/Ulip/CRMP protein 4b; tuc-4; Ulip; unc-33-like phosphoprotein 1; unnamed protein product
Gene Aliases: CRMP-4; CRMP4; DRP-3; DRP3; LCRMP; TUC-4b; TUC4; ULIP; ULIP-1; Ulip1
UniProt ID: (Human) Q14195, (Mouse) Q62188, (Rat) Q62952
Entrez Gene ID: (Human) 1809, (Mouse) 22240, (Rat) 25418
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support