Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
SFRS6 is involved in mRNA splicing and may play a role in the determination of alternative splicing. It belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12. The protein encoded by this gene is involved in mRNA splicing and may play a role in the determination of alternative splicing. The encoded nuclear protein belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 91; MGC5045; pre-mRNA-splicing factor SRP55; splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 6; unnamed protein product
Gene Aliases: 1210001E11Rik; B52; HEL-S-91; SFRS6; SRP55
UniProt ID: (Human) Q13247, (Rat) G3V6S8, (Mouse) Q3TWW8
Entrez Gene ID: (Human) 6431, (Rat) 362264, (Mouse) 67996
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support