Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This gene encodes a member of the Tubby family of bipartite transcription factors. The encoded protein may play a role in obesity and sensorineural degradation. The crystal structure has been determined for a similar protein in mouse, and it functions as a membrane-bound transcription regulator that translocates to the nucleus in response to phosphoinositide hydrolysis. Two transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: candidate tub gene; similar to brain putative tub gene product, GenBank Accession Number U52433; similar to CAEEL48.2K protein, Swiss-Prot Accession Number Q09306; similar to mouse p46 protein. Swiss-Prot Accession Number P46686; first ATG in open reading ; candidate tub gene; similar to C.elegans 48.2 protein Swiss-Prot Accession Number Q09306; similar to mouse p46 protein Swiss-Prot Accession Number P46686; retinal degeneration 5; tubby bipartite transcription factor; tubby candidate; tubby homolog; tubby homologue; unnamed protein product
Gene Aliases: rd5; RDOB; tub
UniProt ID: (Human) P50607, (Mouse) P50586, (Rat) O88808
Entrez Gene ID: (Human) 7275, (Mouse) 22141, (Rat) 25609
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support