Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This gene encodes a tumor suppressor protein that selectively induces apoptosis in cancer cells through intracellular and extracellular mechanisms. The intracellular mechanism involves the inhibition of pro-survival pathways and the activation of Fas-mediated apoptosis, while the extracellular mechanism involves the binding of a secreted form of this protein to glucose regulated protein 78 (GRP78) on the cell surface, which leads to activation of the extrinsic apoptotic pathway. This gene is located on the unstable human chromosomal 12q21 region and is often deleted or mutated different tumors. The encoded protein also plays an important role in the progression of age-related diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: induced in androgen-independent prostate cells by effectors of apoptosis; par-4 induced by effectors of apoptosis; PAR-4/p33; PAR-4/p38; PAWR; PRKC, apoptosis, WT1, regulator; pro-apoptotic protein; prostate apoptosis response 4; prostate apoptosis response 4 protein; Prostate apoptosis response protein 4; prostate apoptosis response protein PAR-4; prostate apoptosis response-4; transcriptional repressor Par-4-like protein PAWR; transcriptional repressor PAR4; unnamed protein product; WT1-interacting protein
Gene Aliases: 2310001G03Rik; Par-4; PAR4
UniProt ID: (Human) Q96IZ0, (Rat) Q62627, (Mouse) Q925B0
Entrez Gene ID: (Human) 5074, (Rat) 64513, (Mouse) 114774
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support