Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
SNAP29 encodes a product that is a member of the SNAP25 gene family, encodes a protein involved in multiple membrane trafficking steps. Two other members of this gene family, SNAP23 and SNAP25, encode proteins that bind a syntaxin protein and mediate synaptic vesicle membrane docking and fusion to the plasma membrane. The protein encoded by this gene binds tightly to multiple syntaxins and is localized to intracellular membrane structures rather than to the plasma membrane. While the protein is mostly membrane-bound, a significant fraction of it is found free in the cytoplasm. Use of multiple polyadenylation sites has been noted for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cerebral dysgenesis, neuropathy, ichthyosis and keratoderma syndrome; FLJ21051; Golgi SNARE; golgi SNARE of 32 kDa; SNAP-29; SNAP29; SNARE; soluble 29 kDa NSF attachment protein; synaptosomal-associated protein, 29kD; synaptosomal-associated protein, 29kDa; synaptosome associated protein 29kDa; syntaxin/SNAP-25 family member; unnamed protein product; vesicle-membrane fusion protein SNAP-29
Gene Aliases: 1300018G05Rik; CEDNIK; Gs32; SNAP-29
UniProt ID: (Human) O95721, (Mouse) Q9ERB0
Entrez Gene ID: (Human) 9342, (Mouse) 67474, (Rat) 116500
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support