Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: ADLAYPVVFA AGLNATHPHQ LMVPGKEGGV PLAPNQPVHG TQADQERLAT CTPSDRTHCA ATPSSSEDTE TVSNSSEGRA SPHDVLETIF VRKVGAFVNK PINQVTLTSL DIPFAMFAPK NLELEDTDPM VNPPDSPETE SPLQGSLHSD GSSGGSSGNT HDDFVMIDFK PAFSKDDILP MDLGTFYREF QNPPQLSSLS
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein ATG1. ATG13 forms a complex with ULK1 and ULK2, the mammalian homologs of ATG1, and with FIP200. This complex is a target of TOR phosphorylation under normal conditions; inhibition of TOR by rapamycin or leucine deprivation leads to dephosphorylation of ATG13, ULK1 and ULK2, which then leads to autophagy. Knockdown of ATG13 inhibits autophagosome formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATG13 autophagy related 13 homolog; FLJ20698; harbinger transposase derived 1; OTTHUMP00000233321; OTTHUMP00000233322; OTTHUMP00000233323; OTTHUMP00000233324; OTTHUMP00000233325; OTTHUMP00000233326; unnamed protein product
Gene Aliases: 1110053A20Rik; D2Ertd391e; Harbi1; Harbi1l; KIAA0652; PARATARG8; RGD1310685
UniProt ID: (Human) O75143, (Mouse) Q91YI1
Entrez Gene ID: (Human) 9776, (Rat) 362164, (Mouse) 51897
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support