Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: KNGSWLYAMC RGIEHDPVKP RQLPKTIGCS KNFPGKTALA TREQVQWWLL QLAQELEERL TKDRNDNDRV ATQLVVSIRV QGDKRLSSLR RCCALTRYDA HKMSHDAFTV IKNCNTSGIQ TEWSPPLTML FLCATKFSAS APSSSTDITS FLS
The XPA gene encodes a zinc metalloprotein that preferentially binds to DNA damaged by UV radiation and chemical carcinogens and is required for the incision step during nucleotide excision repair The XPB and XPD genes encode DNA helicases involved in several DNA metabolic pathways, including DNA repair and transcription, and the XPG gene product is an endonuclease that cuts on the 3' side of a DNA lesion during nucleotide excision repair. Molecular defects in the XP variant (POL H) group maintain normal excision repair, yet they result in a substantial reduction in the ability to synthesize intact daughter DNA strands during DNA replication following DNA damage.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA polymerase eta transcript; FLJ16395; FLJ21978; polymerase (DNA directed), eta; polymerase (DNA) eta; RAD30 homolog A; Saccharomyces cerevisiae RAD30-encoded Pol eta homolog; unnamed protein product; xeroderma pigmentosum variant type protein; XP V
Gene Aliases: RAD30; RAD30A; XP-V; XPV
UniProt ID: (Human) Q9Y253
Entrez Gene ID: (Human) 5429
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support