Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MREPEELMPD SGAVFTFGKS KFAENNPGKF WFKNDVPVHL SCGDEHSAVV TGNNKLYMFG SNNWGQLGLG SKSAISKPTC VKALKPEKVK LAACGRNHTL VSTEGGNVYA TGGNNEGQLG LGDTEERNTF HVISFFTSEH KIKQLSAGSN TSAALTEDGR LFMWGDNSEG QIGLKNVSNV CVPQQVTIGK PVSWISCGYY HSAFVTTDGE LYVFGEPENG KLGLPNQLLG NHRTPQLVSE IPEKVIQVAC GGEHTVVLTE
This gene encodes a protein with a series of six RCC1-like domains (RLDs), characteristic of the highly conserved guanine nucleotide exchange factors. The encoded protein is found in the Golgi body and interacts with RPGRIP1. This protein localizes to the outer segment of rod photoreceptors and is essential for their viability. Mutations in this gene have been associated with X-linked retinitis pigmentosa (XLRP). Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length natures of only some have been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ORF 15; unnamed protein product
Gene Aliases: mRpgr; Rd9; Rp3h
UniProt ID: (Mouse) Q9R0X5
Entrez Gene ID: (Mouse) 19893, (Rat) 367733
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support