Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: YPNASPLLGS SWGGLIHLYT ATARNSYHLQ IHKNGHVDGA PHQTIYSALM IRSEDAGFVV ITGVMSRRYL CMDFRGNIFG SHYFDPENCR FQHQTLENGY DVYHSPQYHF LVSLGRAKRA FLPGMNPPPY SQFLSRRNEI PLIHFNTPIP RRHTRSAEDD SERDPLNVLK PRARMTPAPA SCSQELPSAE DNSPMASDPL GVVRGGRVNT HAGGTGPEGC RPFAKFI
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets, an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia, a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FGF; Fibroblast growth factor; phosphatonin; tumor-derived hypophosphatemia inducing factor
Gene Aliases: ADHR; FGFN; HFTC2; HPDR2; HYPF; PHPTC
UniProt ID: (Human) Q9GZV9
Entrez Gene ID: (Human) 8074
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support