Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSLVAEAFVS QIAAAEPWPE NATLYQQLKG EQILLSDNAA SLAVQAFLQM CNLPIKVVCR ANAEYMSPSG KVPFIHVGNQ VVSELGPIVQ FVKAKGHSLS DGLEEVQKAE MKAYMELVNN MLLTAELYLQ WCDEATVGEI THARYGSPYP WPLNHILAYQ KQWEVKRKMK AIGWGKKTLD QVLEDVDQCC QALSQRLGTQ PYFFNKQPTE LDALVFGHLY TILTTQLTND ELSEKVKNYS NLLAFCRRIE QHYFEDRGKG RLS
The protein encoded by this gene is highly similar to the metaxin 2 protein from mouse, which has been shown to interact with the mitochondrial membrane protein metaxin 1. Because of this similarity, it is thought that the encoded protein is peripherally associated with the cytosolic face of the outer mitochondrial membrane, and that it is involved in the import of proteins into the mitochondrion. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 7.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Metaxin2; mitochondrial outer membrane import complex protein 2; unnamed protein product
Gene Aliases: 1500012G02Rik; MDPS; metaxin-2
UniProt ID: (Human) O75431, (Mouse) O88441
Entrez Gene ID: (Human) 10651, (Mouse) 53375, (Rat) 288150
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support