Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: ECPACYRLVK DKVADHRVKL QELESLIANL GTGDEMVTDQ AFEDRLKEAE REVMDLLREA QDVKDVDQNL MDRLQRVNNT LSSQISRLQN IRNTIEETGN LAEQARAHVE NTERLIEIAS RELEKAKVAA ANVSVTQPES TGDPNNMTLL AEEARKLAER HKQEADDIVR VAKTANDTST EAYNLLLRTL AGENQTAFEI EELNRKYEQA KNISQDLEKQ AARVHEEAKR AGDKAVEIYA SVAQLS
Laminins are essential and abundant structural non-collagenous glycoproteins localizing to basement membranes. Basement membranes (cell-associated extracellular matrices (ECMs)) are polymers of Laminins with stabilizing Type IV Collagen networks, Nidogen and several proteoglycans. Basement membranes are found under epithelial layers, around the endothelium of blood vessels, and surrounding muscle, peripheral nerve and fat cells. Formation of basement membranes influences cell proliferation, phenotype, migration, gene expression and tissue architecture. Each Laminin is a heterotrimer of alpha, beta and gamma chain subunits that undergoes cell-secretion and incorporation into the ECM. Laminins can self-assemble, bind to other matrix macromolecules and have unique and shared cell interactions mediated by Integrins, dystroglycan and cognate Laminin receptors. The human Laminin gamma-1 gene maps to chromosome 1q31 and is ubiquitously expressed in tissues that produce basement membranes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: laminin B2 chain; laminin B2 precursor; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1 subunit gamma; laminin-10 subunit gamma; laminin-11 subunit gamma; laminin-2 subunit gamma; laminin-3 subunit gamma; laminin-4 subunit gamma; laminin-6 subunit gamma; laminin-7 subunit gamma; laminin-8 subunit gamma; laminin-9 subunit gamma; S-LAM gamma; S-laminin subunit gamma; unnamed protein product
Gene Aliases: B2e; LAMB2
UniProt ID: (Human) P11047
Entrez Gene ID: (Human) 3915, (Mouse) 226519, (Rat) 117036
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support