Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
This antibody can also be cross referenced by the term BMS-986012.
The protein encoded by this gene is a Golgi stack membrane protein that is involved in the creation of a precursor of the H antigen, which is required for the final step in the soluble A and B antigen synthesis pathway. This gene is one of two encoding the galactoside 2-L-fucosyltransferase enzyme. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 12FT 2; 12FT2; alpha (1,2) fucosyltransferase; alpha(1,2)FT 2; alpha(1,2)FT2; fucosyltransferase 2 (secretor status included); FUT2 fucosyltransferase 2; galactoside 2-alpha-L-fucosyltransferase 2; galactoside 2-L-fucosyltransferase; GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 2; secretor blood group alpha-2-fucosyltransferase; secretor factor; type 1 galactoside alpha-(1,2)-fucosyltransferase FUT2; type 2 galactoside alpha-(1,2)-fucosyltransferase FUT2; unnamed protein product
Gene Aliases: B12QTL1; SE; Se2; SEC2; sej
UniProt ID: (Human) Q10981
Entrez Gene ID: (Human) 2524
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support