Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
This antibody can also be cross referenced by the term NG-101.
Nogo-B is a novel myelin-associated, intermediate-length splice variant of Nogo belonging to the reticulon family. It is a multidomain neurite growth inhibitory protein containing several discrete regions with growth inhibitory functions and is a component of the CNS myelin that prevents axonal regeneration in the adult vertebrate CNS. Nogo-B along with NGBR mediated chemotaxis in HUVECs and induced tube formation in 3-dimensional cultures. It is mostly found in intact blood vessels, smooth muscle cells, and endothelial cell, with its N terminus oriented extracellularly and is a regulator of vascular homeostasis and remodeling. It plays an important role in sporadic and familial amyotrophic lateral sclerosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ASY; foocen; Human NogoA; MGC116054; My043 protein; neurite growth inhibitor 220; neurite outgrowth inhibitor; neuroendocrine-specific protein C homolog; NOGO-A; RTN4-A; NOGO-B; RTN4-B1; NOGO-C; RTN4-C; reticulon; reticulon 5; RTN4-Aa; RTN4-Ab; RTN4-B2; RTN4-D; RTN4-E; RTN4-F; RTN4-G; RTN4a; RTN4b; RTN4c; unnamed protein product
Gene Aliases: ASY; Nbla00271; Nbla10545; NI220/250; NOGO; NOGOA; NOGOB; NSP; NSP-CL; RTN-X; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C
UniProt ID: (Human) Q9NQC3
Entrez Gene ID: (Human) 57142
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support