Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
MA5-60133 is a human/mouse chimeric antibody. The variable regions from one species have been grafted to the backbone of the other species.
The CD3 antibody reacts with the intracytoplasmic portion of the CD3 antigen expressed by normal and neoplastic T-lymphocytes. It stains human T-lymphocytes in both cortex and medulla of the thymus and in peripheral lymphoid tissues. CD20 is a non-Ig differentiation antigen of B-lymphocytes and the expression of CD20 is restricted to normal and neoplastic B-lymphocytes, being absent from all other leukocytes and tissues. The CD3-CD20 cocktail reveals lymphoid architecture in neoplastic situations.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-lymphocyte cell-surface antigen B1; CD20 antigen; CD20 receptor; CD3 antigen, delta subunit; CD3 delta; CD3-epsilon; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule, delta (CD3-TCR complex); CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule, epsilon (CD3-TCR complex); CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); leukocyte surface antigen Leu-16; membrane-spanning 4-domains, subfamily A, member 1; OKT3, delta chain; T-cell antigen receptor complex, epsilon subunit of T3; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; T-cell surface antigen T3/Leu-4 epsilon chain; unnamed protein product
Gene Aliases: B1; Bp35; CD20; CD3-DELTA; CD3-GAMMA; CD3DELTA; CD3epsilon; CD3GAMMA; CVID5; FMC7; IMD17; IMD18; IMD19; LEU-16; S7; T3D; T3E; T3G; TCRE
UniProt ID: (Human) P04234, (Human) P07766, (Human) P09693, (Human) P11836
Entrez Gene ID: (Human) 915, (Human) 916, (Human) 917, (Human) 931
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support