Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SARS-CoV-2 Spike Protein RBD Omicron Antibodies

          Invitrogen

          SARS-CoV-2 Spike Protein RBD Omicron (BA.4/BA.5/BA.2.75) Chimeric Recombinant Human Monoclonal Antibody (D003)

          View all (4) SARS-CoV-2 Spike Protein RBD Omicron antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SARS-CoV-2 Spike Protein RBD Omicron (BA.4/BA.5/BA.2.75) Chimeric Recombinant Human Monoclonal Antibody (D003)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}

          Product Details

          MA5-60601

          Applications
          Tested Dilution
          Publications

          ELISA (ELISA)

          0.1-0.2 µg/mL
          -

          Neutralization (Neu)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Virus

          Host/Isotype

          Human / IgG1, kappa

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          D003

          Immunogen

          Recombinant SARS-CoV-2 Omicron BA.4/BA.5 Spike ECD Protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          20 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3701224

          Product Specific Information

          Specifically recognize the SARS-CoV-2 Omicron BA.4/BA.5/BA.5.2 Spike RBD and ECD (Can block the binding between RBD and ACE2). No cross-reactivity with SARS-CoV-2 wild type, Alpha, Beta, Delta variant. Has Medium cross-reactivity in ELISA with SARS-CoV-2 Omicron BA.2.75 Spike RBD.. Has cross-reactivity in ELISA with: SARS-CoV-2 Omicron (BQ.1.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike RBD Protein; SARS-CoV-2 Omicron (BF.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4.6/BF.7) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.4.6) Spike S1+S2 trimer Protein; No cross-reactivity in ELISA with: SARS-CoV-2 Omicron (XBB.1.16) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike RBD Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.3.20) Spike RBD Protein; SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein; SARS-CoV-2 Omicron (B.1.1.529) Spike RBD Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike RBD Protein; SARS-CoV-2 Beta (B.1.351) Spike S1+S2 trimer Protein; SARS-CoV-2 Beta (B.1.351, B.1.351.2, B.1.351.3) Spike RBD Protein; SARS-CoV-2 Delta (B.1.617.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Delta (B.1.617.2) Spike RBD Protein; SARS-CoV-2 Spike S1+S2 (D614G) trimer Protein; SARS-CoV-2 Spike RBD Protein

          Target Information

          The severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) variant Omicron (B|1|1|529) has attracted great concerns since its identification in South Africa|Omicron is the fifth variant of concern (VOC) after Alpha (B|1|1|7), Beta (B|1|351), Gamma (P|1) and Delta (B|1|617|2)|Omicron variant harbors >30 substitutions/deletions/insertions in the spike, including 15 substitutions in receptor-binding domain (RBD) [PMID: 35397623]

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 2019-nCoV Spike; COVID-19 Spike; SARS-CoV-2 Spike

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline