Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute at 0.5 mg/mL in sterile PBS.
Galectin-12, also designated galectin-related inhibitor of proliferation 1 in mouse, is a 314 amino acid protein encoded by the human gene LGALS12. Galectin-12 is a member of the galectin family consisting of b-galactoside-binding proteins with conserved carbohydrate recognition domains. Galectin-12 binds lactose and may participate in the apoptosis of adipocytes. This protein is preferentially expressed in peripheral blood leukocytes and adipocytes. Galectin-12 is induced by cell cycle block at the G1 phase and causes G1 arrest when overexpressed. The galectin-12 gene is expressed in mouse preadipo-cytes and is upregulated when preadipocytes undergo cell cycle arrest, concomitant with acquisition of the competence to undergo differentiation in response to adipogenic hormone stimulation. Galectin-12 is an adipocyte-expressed protein which is downregulated by various insulin resistance-inducing hormones. As a result, galectin-12 may play a role in the pathogenesis of insulin resistance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: galectin-related inhibitor of proliferation; GRIP1a; GRIP1b; lectin, galactoside-binding, soluble, 12; testicular secretory protein Li 26; unnamed protein product
Gene Aliases: GAL12; GRIP1
UniProt ID: (Human) Q96DT0
Entrez Gene ID: (Human) 85329
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support