Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute at 0.5 mg/mL in sterile PBS.
The inter-alpha-trypsin inhibitors (ITI) are a family of structurally related plasma serine protease inhibitors involved in extracellular matrix stabilization and in prevention of tumor metastasis. The ITI family contains multiple proteins made up of a light chain (see MIM 176870) and a variable number of heavy chains (Salier et al., 1987 [PubMed 2446322]; Himmelfarb et al., 2004 [PubMed 14744536]). ITI-HC2 may act as a carrier of hyaluronan in serum or as a binding protein between hyaluronan and other matrix protein, including those on cell surfaces in tissues to regulate the localization, synthesis and degradation of hyaluronan which are essential to cells undergoing biological processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HC2; inter-alpha (globulin) inhibitor H2; inter-alpha (globulin) inhibitor, H2 polypeptide; inter-alpha-inhibitor heavy chain 2; Inter-alpha-trypsin inhibitor complex component II; inter-alpha-trypsin inhibitor heavy chain; Inter-alpha-trypsin inhibitor heavy chain H2; inter-alpha-trypsin inhibitor heavy chain old gene name 'ITI'; ITI heavy chain H2; PSMA1; Serum-derived hyaluronan-associated protein; SHAP; unnamed protein product
Gene Aliases: H2P; IGHEP2; ITI-HC2; ITIH2; SHAP
UniProt ID: (Human) P19823
Entrez Gene ID: (Human) 3698
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support