Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute at 0.5 mg/mL in sterile PBS.
OMD may be implicated in biomineralization processes. This protein has a function in binding of osteoblasts via the alpha(V)beta(3)-integrin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Keratan sulfate proteoglycan osteomodulin; KSPG osteomodulin; OSAD; Osteoadherin; Osteomodulin; UNQ190/PRO216SLRR2C
Gene Aliases: Omd; OSAD; SLRR2C
UniProt ID: (Mouse) O35103
Entrez Gene ID: (Mouse) 27047
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support