Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute at 0.5 mg/mL in sterile PBS.
Hyaluronan and proteoglycan link protein 4 (HAPLN4), also known as Brain Link Protein 4 (BRAL1), is a gene located on chromosome 5q14.2, encoding a member of the link protein family that plays a crucial role in stabilizing interactions between hyaluronan and proteoglycans within the extracellular matrix. HAPLN4 is predominantly expressed in the brain, especially within the central nervous system where it contributes to the structural integrity and functional modulation of neural tissues. The protein is key in maintaining the organization of the extracellular matrix by binding hyaluronan and other matrix components, facilitating cell adhesion and signaling. This is vital for processes like neuronal migration, synapse formation, and neuronal differentiation during brain development. Variations in HAPLN4 expression or function are associated with neural developmental disorders and may influence the progression of neurodegenerative diseases due to its critical role in preserving the extracellular matrix architecture and neural connectivity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain link protein 2; Hyaluronan and proteoglycan link protein 4; link protein
Gene Aliases: BRAL2; HAPLN4; KIAA1926
UniProt ID: (Human) Q86UW8
Entrez Gene ID: (Human) 404037
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support