Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Reconstitute at 0.5 mg/mL in sterile PBS.
CADM4 is involved in the cell-cell adhesion. Has calcium- and magnesium-independent cell-cell adhesion activity. May have tumor-suppressor activity. Fukuhara H., Oncogene 20:5401-5407(2001). Grimwood J., Nature 428:529-535(2004)0. Williams Y.N., Oncogene 25:1446-1453(2006).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: immunoglobulin superfamily member 4C; immunoglobulin superfamily, member 4C; membrane glycoprotein Tsll2; NECL-4; nectin-like 4; nectin-like protein 4; SynCAM 4; TSLC1-like 2; TSLC1-like protein 2; TSLL2; TSLL2; transmembrane protein; unnamed protein product; urokinase plasminogen activator receptor
Gene Aliases: Igdf4c; IGSF4C; Necl-4; NECL4; synCAM4; TSLL2; Upar
UniProt ID: (Human) Q8NFZ8, (Rat) Q1WIM1, (Mouse) Q8R464
Entrez Gene ID: (Human) 199731, (Rat) 365216, (Mouse) 260299
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support