Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute at 0.5 mg/mL in sterile PBS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 1 protein; expressed in non-metastatic cells 1 protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 1, protein; expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; Farnesyl diphosphate kinase NME1; GAAD; granzyme A-activated DNase; HEL-S-155an; Histidine protein kinase NDK; Histidine protein kinase NDKB; Histidine protein kinase NME1; Metastasis inhibition factor NM23; NDK A; NDK B; NDK1; NDK2; NDP kinase A; NDP kinase alpha; NDP kinase B; NDP kinase beta; NDPK-A; NDPK2; NM23-H1; nm23-H2; nm23-H2S product (putative NDP kinase); putative; nm23-M1; nm23-M2; NM23H1; NME1; NME2; non-metastatic cells 1, protein (NM23A) expressed in; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; Non-specific serine/threonine protein kinase NME1; Nucleoside diphoshate kinase 1; Nucleoside diphoshate kinase 2; Nucleoside diphosphate kinase A; Nucleoside diphosphate kinase B; Nucleoside diphosphate kinases 2; nucleoside-diphosphate kinase 1; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase; nucleotide diphosphate kinase B; nucloside diphosphate kinase; P18; Putative 3'-5'-DNA exonuclease NDK1; Putative granzyme A-activated DNA endonuclease; Tumor metastatic process-associated protein; unnamed protein product
Gene Aliases: AL024257; AWD; GAAD; NB; NBS; NDK2; NDKA; NDKB; NDPK-A; NDPK-B; NDPKA; NDPKB; NM23; nm23-2; NM23-H1; NM23-H2; NM23-M1; nm23-M2; NM23A; NM23B; NME1; NME2; p18-12d; PUF
UniProt ID: (Human) P15531, (Human) P22392, (Mouse) P15532, (Rat) Q05982, (Rat) P19804, (Mouse) Q01768
Entrez Gene ID: (Human) 4830, (Human) 4831, (Mouse) 18102, (Rat) 191575, (Rat) 83782, (Mouse) 18103
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support