Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
This gene encodes a member of the SH2-domain containing mediators family. The encoded protein mediates activation of various kinases and may function in cytokine and growth factor receptor signaling and cellular transformation. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp547G1110; FLJ30542; Pro-rich, PH and SH2 domain-containing signaling mediator; PSM; SH2 B; SH2 domain-containing protein 1B; SH2 domain-containing putative adapter SH2-B; SH2-B signaling protein; SH2B adapter protein 1; SH2B1 protein; unnamed protein product
Gene Aliases: KIAA1299; PSM; SH2B; SH2B1
UniProt ID: (Human) Q9NRF2
Entrez Gene ID: (Human) 25970
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support