Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute at 0.5 mg/mL in sterile PBS.
Together with the tyrosine-protein kinase SYK, enhances production of the cytokine CXCL8/IL-8 via the NFKB pathway and may thus have a role in the intestinal immune response.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: carcinoembryonic antigen related cell adhesion molecule 20; Carcinoembryonic antigen-related cell adhesion molecule 20; CEA cell adhesion molecule 20; CEACAM20-4L; CEACAM20-4S; CEACAM20-5L; CEACAM20-5S; Cell adhesion molecule CEACAM20; GPAD9366
Gene Aliases: CEACAM20; UNQ9366; UNQ9366/PRO34155
UniProt ID: (Human) Q6UY09
Entrez Gene ID: (Human) 125931
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support