Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 18
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Positive Control: Saos-2 cell lysates, human osteosarcoma tissue, mouse embryo tissue, rat embryo tissue, E14.5 mouse embryo tissue.
Tissue Specificity: Not detected.
Subcellular Location: Nucleus.
Sequence Similarities: 95% Mouse.
Predicted band size: 45 kDa.
SP7 is a C2H2-type zinc finger transcription factor of the SP gene family and a putative master regulator of bone cell differentiation (Gao et al., 2004 [PubMed 15474293]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C2H2 zinc finger transcription factor; OSX; MGC126598; osteoblast transcription factor; osterix; transcription factor; transcription factor containing 3 Cys2His2-type zinc fingers; Sp7; unnamed protein product; zinc finger protein; zinc finger protein osterix
Gene Aliases: 6430578P22Rik; C22; OI11; OI12; osterix; OSX
UniProt ID: (Human) Q8TDD2, (Mouse) Q8VI67
Entrez Gene ID: (Human) 121340, (Rat) 300260, (Mouse) 170574
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support