Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Synapsin 1/2 Antibodies

          Invitrogen

          Synapsin 1/2 Recombinant Rabbit Monoclonal Antibody (PSH10-29)

          Advanced Verification

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Synapsin 1/2 Recombinant Rabbit Monoclonal Antibody (PSH10-29)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (26)
          • Advanced Verification (5)
          Synapsin 1/2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Synapsin 1/2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 31

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Synapsin 1/2 Antibody (MA562922) in ICC/IF

          Immunofluorescence analysis of Synapsin 1/2 was performed using IMR-32 cells. The cells were fixed with 4% paraformaldehyde for 5 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with Synapsin 1/2 Recombinant Rabbit Monoclonal Antibody (PSH10-29) (Product # MA562922) at 1:100 in 0.1% BSA, incubated at 4 degree celsiu... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Synapsin 1/2 Antibody in Immunocytochemistry (ICC/IF)
          Synapsin 1/2 Antibody in Immunocytochemistry (ICC/IF)
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Synapsin 1/2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Synapsin 1/2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Synapsin 1/2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Synapsin 1/2 Antibody in Western Blot (WB)
          Synapsin 1/2 Antibody in Western Blot (WB)
          Synapsin 1/2 Antibody in Western Blot (WB)
          Synapsin 1/2 Antibody in Western Blot (WB)
          Synapsin 1/2 Antibody in Western Blot (WB)
          Synapsin 1/2 Antibody in Immunoprecipitation (IP)
          Synapsin 1/2 Antibody
          Synapsin 1/2 Antibody
          Synapsin 1/2 Antibody
          Synapsin 1/2 Antibody
          Synapsin 1/2 Antibody
          Synapsin 1/2 Recombinant Rabbit Monoclonal Antibody (PSH10-29)

          Product Details

          MA562922

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:1,000
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:500
          -

          Immunocytochemistry (ICC/IF)

          1:500
          -

          Immunoprecipitation (IP)

          1-2 µg per sample
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          PSH10-29

          Immunogen

          Recombinant protein within human Synapsin I aa 1-705.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          20 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 0.1% BSA, 40% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Positive Control: Mouse brain tissue lysate, Rat brain tissue lysate, mouse primary neuronal, human brain tissue, mouse brain tissue, mouse retina tissue, rat brain tissue, rat retina tissue.

          Tissue Specificity: Tissue enriched (brain)

          Subcellular Location: Synapse, Golgi apparatus, Presynapse, Cytoplasmic vesicle, secretory vesicle, synaptic vesicle.

          Sequence Similarities: 95% Mouse,Rat

          Predicted band size: 62 kDa.

          Target Information

          The SYN1 gene, located on chromosome Xp11.3-p11.23, encodes synapsin I, a neuronal phosphoprotein critical for the regulation of synaptic vesicle release and neurotransmitter mediation. Synapsin I is involved in synaptic plasticity and the maintenance of synaptic vesicle pools, modulating neurotransmission and synaptic strength. The SYN2 gene, located on chromosome 3p25-p26, encodes synapsin II, another protein that plays a similar role in synaptic vesicle trafficking and neurotransmitter release. Both synapsin I and II are essential for proper neuronal function and are highly expressed in the brain. Dysregulations or mutations in these genes can be associated with neurological disorders such as epilepsy and schizophrenia, indicating their importance in maintaining the integrity of neural networks.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Brain protein 4.1; synapsin 1; synapsin 2; Synapsin I; synapsin Ib; Synapsin II; Synapsin-1; Synapsin-2; synaptic vesicle protein; truncated synapsin 1-S; unnamed protein product

          View more View less

          Gene Aliases: 2900074L19Rik; AI836018; AI841723; EPILX; EPILX1; MRX50; Syn-1; SYN1; Syn1-S; SYN1a; SYN1b; SYN2; SYNI; SYNII

          View more View less

          UniProt ID: (Human) P17600, (Human) Q92777, (Rat) P09951, (Mouse) O88935, (Mouse) Q64332, (Rat) Q63537

          View more View less

          Entrez Gene ID: (Human) 6853, (Human) 6854, (Rat) 24949, (Mouse) 20964, (Mouse) 20965, (Rat) 29179

          View more View less

          Function(s)
          actin binding protein binding ATP binding protein kinase binding identical protein binding calcium-dependent protein binding microtubule cortical anchor activity catalytic activity protein dimerization activity cytoskeletal protein binding membrane trafficking regulatory protein
          Process(es)
          chemical synaptic transmission neurotransmitter secretion regulation of neurotransmitter secretion neuron development modulation of synaptic transmission synapse organization synaptic vesicle clustering regulation of synaptic vesicle cycle synaptic vesicle cycle regulation of synaptic vesicle exocytosis calcium ion regulated exocytosis metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline