Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: ALPAQEQLRP KPQLEQENIK SSSPPSSDFA KEEVIGMVTR AHKDTFMYNQ EQQENSAESM QPQRGERIPK NMEQYNLNHD H
This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EAR4; HZF2; orphan nuclear receptor HZF-2; orphan nuclear receptor RVR; orphan receptor; Rev-erb beta; rev-erb-beta; Rev-erb-Beta2; Rev-erba beta; Rev-ErbA-beta; Rev-erbabeta; unnamed protein product
Gene Aliases: HZF-2; Rev-erb; RVR
UniProt ID: (Mouse) Q60674, (Rat) Q63504
Entrez Gene ID: (Mouse) 353187, (Rat) 259241
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support