Metrios 6S (S)TEM
Metrios 6S (S)TEM
Thermo Scientific™
Metrios 6S (S)TEM
The Thermo Scientific Metrios 6S (S)TEM enables automated TEM metrology utilizing automated TEM grid handling and seamless data connectivity between systems within the Thermo Scientific AutoLab Solution. Leveraging our expertise in these areas, the Metrios 6S (S)TEM is designed to address the distinct challenges encountered in scaling lab operations.
Have Questions?
Número de catálogo METRIOS-6S-STEM
Precio (MXN)
-
Solicite un presupuesto
The Thermo Scientific Metrios 6S (S)TEM enables automated TEM metrology utilizing automated TEM grid handling and seamless data connectivity between systems within the Thermo Scientific AutoLab Solution. Leveraging our expertise in these areas, the Metrios 6S (S)TEM is designed to address the distinct challenges encountered in scaling lab operations.
Fully automated semiconductor metrology
The Metrios 6S (S)TEM is a new-generation, fully automated instrument that enables high-volume TEM metrology. The Metrios 6S (S)TEM features newly designed hardware and machine-learning-based capabilities. As part of the AutoLab Solution, SEMI standard compliant TEM grid handling and host connection are seamlessly integrated, allowing automated TEM grid transfer from the Helios 6 HXS FIB-SEM and continuous data connectivity with the lab MES system. Along with Smart Automation software, this combination provides improved TEM metrology productivity and recipe-free automation for scalable lab operations and resource optimization.
Automated TEM lamella handling
The new Metrios EFEM (Equipment Front End Module) is an innovative TEM lamella handling system designed for the Metrios 6S (S)TEM as part of the AutoLab Solution. It provides an automated, operator-free solution for high-throughput loading and unloading of TEM lamella, seamlessly integrating with automated materials handling systems (AMHS) via overhead tracks (OHT) or autonomous mobile robots (AMR). In this workflow, samples are delivered to the Helios 6 HXS (S)TEM for creation of lamellas, which are then transferred to the Metrios 6S (S)TEM for high-precision metrology data collection.
Automated data connectivity for TEM metrology
Automated data connectivity facilitates seamless SEMI standard communication between your manufacturing execution system (MES) and the Metrios 6S (S)TEM’s workflow control software, including interfacing with the OHT or AMR material handling system. If the Helios 6HXS and the Metrios 6S (S)TEMs are not connected to the MES system, they can be linked directly for workflow data connectivity. The workflow control software is used to create and schedule imaging jobs for the Metrios 6S (S)TEM. These jobs are then executed in the Metrios 6S (S)TEM’s application software. This enables a streamlined workflow with enhanced data connectivity and automated processing.
Automated TEM imaging and metrology
The Metrios 6S (S)TEM offers sophisticated, automated metrology, delivering precise nanoscale measurements through automated image acquisition and analysis, essential for semiconductor manufacturing. Its Smart Automation software significantly reduces set-up time for new processes and sample types, ensuring ease of use and scalability by leveraging advanced machine-learning algorithms. The instrument’s flexibility allows operation in both full-auto and semi-auto modes, adapting to specific lab needs and accommodating a wide range of challenging semiconductor device types. For model training, a new web-based segmenter and object detection algorithm accelerate the time to build robust Al models. For additional flexibility, conventional recipe-based automation is also available.
Metrios 6S (S)TEM technical highlights
TEM grid handling
SEMI standard-compliant, operator-free grid handling via AMHS.
Data connectivity
SEMI standard-compliant software interface for connection to MES.
Metrology
TEM and STEM mode metrology accuracy: Less than 0.75% combined error in distortion and magnification calibration.
Application software
Smart Automation software allows setup of new automation routines in <4 hours for select workflows.
EDS quantification using Thermo Scientific Velox Software, featuring dynamic correction of holder shadowing as a function of tilt.
EDS
Ultra-X detector provides high-sensitivity, windowless EDS detection with high solid angle and high cleanliness.
Electron column
Three-lens condenser system with indicators for convergence angle and size of the illuminated area, allowing for quantitative measurement of electron dose and illumination conditions.
The S-CORR probe corrector provides sub-Ångström imaging resolution at 60 kV as specification and an order of magnitude improvement in optical stability.
The S-CORR corrector simultaneously corrects A5 for all accelerating voltages. Flexible, high-tension range of 60–200 kV.
Choice of ultra-stable, high-brightness Schottky field emission gun (X-FEG) or ultra-high- brightness cold field emission gun (X-CFEG) with an energy resolution of <0.4 eV.
L6
Stage
Five-axis, piezo-driven Smart Stage with minimum XYZ step size of ≤20 pm.
Holder
High-accuracy, double-tilt holder designed for the Smart Stage.
Alpha-tilt, under-goniometer software control; Alpha tilt range ±40 degrees for high- accuracy, double-tilt holder.
Learn more about Metrios 6S (S)TEM
Especificaciones
TipoSTEM Microscope
Unit SizeEach