MicroAmp™ 8-Tube Strip, 0.2 mL - FAQs
MicroAmp™ 8-Tube Strip, 0.2 mL - FAQs
View additional product information for MicroAmp™ 8-Tube Strip, 0.2 mL - FAQs (N8010580)
1 product FAQs found
What is the shelf life of MicroAmp Optical 8-tube Strip, 0.2 ml (Cat. No. N8010580)?
Our plastics have an expiry date of 5 years from the manufacturing date. The exact expiry date can be found in the CofA.