GeneAmp™ dNTP Blend (100 mM), 1 mL - FAQs
GeneAmp™ dNTP Blend (100 mM), 1 mL - FAQs
View additional product information for GeneAmp™ dNTP Blend (100 mM) - FAQs (N8080261)
1 product FAQs found
Why do some dNTP mixes contain dTTP and others TTP?
Thymine is normally found as a deoxyribose. Thus, TTP and dTTP are essentially the same thing. Both types of mixes can be used in PCR reactions