Ovalbumin, Texas Red™ Conjugate
Invitrogen™

Ovalbumin, Texas Red™ Conjugate

The red fluorescent Texas Red ovalbumin conjugate is a protein with a relatively low molecular weight (∼45,000 daltons) labeled with深入閱讀
Have Questions?
產品號碼Quantity
O230215 mg
產品號碼 O23021
價格 (TWD)
5,160.00
線上優惠
Ends: 31-Dec-2026
7,360.00
您節省 2,200.00 (30%)
Each
Quantity:
5 mg
價格 (TWD)
5,160.00
線上優惠
Ends: 31-Dec-2026
7,360.00
您節省 2,200.00 (30%)
Each
The red fluorescent Texas Red ovalbumin conjugate is a protein with a relatively low molecular weight (∼45,000 daltons) labeled with the bright, photostable and pH-insensitive Texas Red dye.
For Research Use Only. Not for use in diagnostic procedures.
規格
Label TypeClassic Dyes
Product LineTexas Red
Protein SubtypeAlbumin Proteins
Quantity5 mg
Shipping ConditionRoom Temperature
ConjugateTexas Red
FormLyophilized
Unit SizeEach
內容物與存放
Store in freezer (-5 to -30°C) and protect from light.

引用資料與參考文獻 (25)

引用資料與參考文獻
Abstract
A prelysosomal compartment sequesters membrane-impermeant fluorescent dyes from the cytoplasmic matrix of J774 macrophages.
Authors:Steinberg TH, Swanson JA, Silverstein SC
Journal:J Cell Biol
PubMed ID:3417786
'After the membrane impermeant dye Lucifer Yellow is introduced into the cytoplasmic matrix of J774 cells, the dye is sequestered within cytoplasmic vacuoles and secreted into the extracellular medium. In the present work we studied the intracellular transport of Lucifer Yellow in J774 macrophages and the nature of the cytoplasmic ... More
Identification of Legionella pneumophila mutants that have aberrant intracellular fates.
Authors:Swanson MS, Isberg RR
Journal:Infect Immun
PubMed ID:8698483
'After uptake by macrophages, Legionella pneumiophila evades phagosome-lysosome fusion and replicates in a compartment associated with the endoplasmic reticulum. A collection of bacterial mutants defective for growth in macrophages were isolated, and the intracellular fate of each mutant strain was analyzed by fluorescence microscopy. To measure intracellular replication, bacteria inside ... More
Involvement of early and late lysosomes in the degradation of mannosylated ligands by rat liver endothelial cells.
Authors:Kjeken R, Brech A, Løvdal T, Roos N, Berg T
Journal:Exp Cell Res
PubMed ID:7843273
'The intracellular transport and degradation of endocytosed mannosylated albumin (Man-BSA) was studied in cell cultures of rat liver endothelial cells by subcellular fractionation, fluorescence microscopy, and electron microscopy. The ligand used for subcellular fractionation experiments was labeled with 125I-labeled tyramine cellobiose or 131I-labeled tyramine cellobiose. The labeled degradation products are ... More
Analysis of the intracellular fate of Legionella pneumophila mutants.
Authors:Swanson MS, Isberg RR
Journal:Ann N Y Acad Sci
PubMed ID:8993346
'L. pneumophila is a model organism for investigating the mechanisms by which intracellular pathogens acquire the metabolites needed for replication while evading the microbicidal mechanisms of the macrophage. We determined that intracellular L. pneumophila replicate in close association with the endoplasmic reticulum and suggest that L. pneumophila exploits the macrophage ... More
Fluorescent labeling of endocytic compartments.
Authors:Swanson J
Journal:Methods Cell Biol
PubMed ID:2464119