Oregon Green™ 488 BAPTA-1 dextran, Potassium Salt, 10,000 MW, Anionic
Invitrogen™

Oregon Green™ 488 BAPTA-1 dextran, Potassium Salt, 10,000 MW, Anionic

Dextran forms of these calcium indicators show dramatic reduction in both leakage and compartmentalization compared to AM ester forms

Have Questions?
Catalog NumberQuantity
O67985 mg
Catalog number O6798
Price (USD)
626.00
Each
Quantity:
5 mg
Price (USD)
626.00
Each

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues. Cells may be physically loaded with the cell-impermeant salt forms of these dextran-conjugated indicators using patch pipette, microinjection, or our Influx pinocyte-loading reagent (Cat. No. I14402). The fluorescence signal from these cells is measured using fluorescence microscopy. The dextran forms of our calcium indicators show a dramatic reduction in both leakage and compartmentalization compared to the AM ester forms.

Calcium Indicator (Cell-Impermeant Salts, Dextran Conjugates) Specifications:

  • Label (Ex/Em): Oregon Green™ 488 BAPTA-1 (494/523 nm)
  • Fluorescence intensity increase upon binding Ca2+: ∼14 fold
  • Dextran size: 10,000 MW
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

Spectral Characteristics of Molecular Probes™ Calcium Indicators

These probes are excited by visible light, and because the energy required for excitation is low, the potential for cellular photodamage is reduced. Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of the spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem.

More Choices for Fluorescent Calcium Indicators

We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios, for example dextran versions for reduced leakage and compartmentalization and BAPTA conjugates for detecting high-amplitude calcium transients. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeOregon Green
Emission Wavelength523 nm
Excitation Wavelength494 nm
Quantity5 mg
Shipping ConditionRoom Temperature
For Use With (Application)Calcium Indicator
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, Microplate Reader
Product LineOregon Green
Product TypeReagent
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Citations & References (37)

Citations & References
Abstract
Spatially organised mitochondrial calcium uptake through a novel pathway in chick neurones.
Authors:Coatesworth W, Bolsover S
Journal:Cell Calcium
PubMed ID:16338004
'A brief depolarisation of chick sensory neurones evokes a calcium increase in mitochondria that peaks 1-2s after the depolarisation event and then decays over tens of seconds. Peripheral mitochondria take up more calcium than do central ones, even when the cytosolic calcium increase is spatially homogeneous. The calcium influx into ... More
The sources and sequestration of Ca(2+) contributing to neuroeffector Ca(2+) transients in the mouse vas deferens.
Authors:Brain KL, Cuprian AM, Williams DJ, Cunnane TC
Journal:J Physiol
PubMed ID:14500773
'The detection of focal Ca(2+) transients (called neuroeffector Ca(2+) transients, or NCTs) in smooth muscle of the mouse isolated vas deferens has been used to detect the packeted release of ATP from nerve terminal varicosities acting at postjunctional P2X receptors. The present study investigates the sources and sequestration of Ca(2+) ... More
Control of IP(3)-mediated Ca2+ puffs in Xenopus laevis oocytes by the Ca2+-binding protein parvalbumin.
Authors:John LM, Mosquera-Caro M, Camacho P, Lechleiter JD
Journal:J Physiol
PubMed ID:11507154
'1. Elementary events of Ca2+ release (Ca2+ puffs) can be elicited from discrete clusters of inositol 1,4,5 trisphosphate receptors (IP(3)Rs) at low concentrations of IP(3). Ca(2+) puffs have rarely been observed unless elicited by either hormone treatment or introduction of IP(3) into the cell. However, cells appear to have sufficient ... More
Calcium in sympathetic varicosities of mouse vas deferens during facilitation, augmentation and autoinhibition.
Authors:Brain KL, Bennett MR
Journal:J Physiol
PubMed ID:9279805
'1. The sympathetic nerve terminals of the mouse vas deferens were loaded with the calcium indicator Oregon Green 488 BAPTA-1 by orthograde transport along the postganglionic nerves. Changes in the calcium concentration in the varicosity (delta [Ca2+]v) were determined following single impulses, and short (5-impulse) and long (200-impulse) trains at ... More
Coupling of calcium homeostasis to axonal sodium in axons of mouse optic nerve.
Authors:Verbny Y, Zhang CL, Chiu SY
Journal:J Neurophysiol
PubMed ID:12163532
'Axonal populations in neonatal and mature optic nerves were selectively stained with calcium dyes for analysis of calcium homeostasis and its possible coupling to axonal Na. Repetitive nerve stimulation causes a rise in axonal [Ca(2+)](i) the posttetanus recovery of which is impeded by increasing the number of action potentials in ... More