Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 9
Suggested positive control: human breast cancer and kidney tissue.
The trace metal copper (Cu) plays a crucial role in mammalian cells as a cofactor for many enzymes. Cu-related genetic diseases, such as Menkes disease and Wilson disease, arise from a lack of Cu homeostasis in mammalian cells. CTR1 is a high-affinity copper-uptake protein. The C-terminal domain is similar to the Raf family of protein kinases, but it's first two-thirds encodes a novel protein domain. CTR1 provides the primary avenue for copper uptake in mammalian cells, thereby, affecting Cu homeostasis and embryonic development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: At5g03730; AtCTR1; CONSTITUTIVE TRIPLE RESPONSE 1; copper transport 1 homolog; Copper transporter 1; Copper uptake transporter 1; CTR1; F17C15.150; F17C15_150; hCTR1; high affinity copper uptake protein; High affinity copper uptake protein 1; high-affinity copper uptake protein; Liver regeneration-related protein LRRGT00200; rCTR1; SIS1; solute carrier family 13 (sodium/sulphate symporters), member 1; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporters), member 1; Solute carrier family 31 member 1; SUGAR-INSENSITIVE 1; unnamed protein product
Gene Aliases: 4930445G01Rik; AI787263; AU016967; COPT1; ctr-1; CTR1; LRRGT00200; NSCT; SLC31A1; Xctr1; XELAEV_18038194mg; zgc:76839
UniProt ID: (Human) O15431, (Pig) Q8WNR0, (Rat) Q9JK41, (Mouse) Q8K211
Entrez Gene ID: (Human) 1317, (Pig) 397186, (Dog) 481678, (Rat) 171135, (Mouse) 20529, (Zebrafish) 403028, (Xenopus) 379422
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support