Hamburger Menu Button
Thermo Fisher Scientific Logo
로그인
회원이 아니신가요? 계정 생성하기
  • 모든 제품 주문
    • 항체
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR 분석
    • 피펫 및 피펫 팁
    • 실험실용 원심분리기
    • 초저온 냉동고
    • 분광학
    • 비이커
    • PCR 장비 및 용품
    • 제품 카테고리 전체보기
  • 응용 분야 및 기법
    • 세포 분석
    • 실험실 장비
    • Real-Time PCR
    • PCR
    • 크로마토그래피
    • 세포 배양 및 트랜스펙션
    • DNA/RNA 추출과 분석
    • 단백질 생물학
    • 유세포분석
    • 화학 제품
    • 응용 분야 및 기법 전체보기
  • 서비스
    • 맞춤형 서비스
    • 엔터프라이즈급 실험실 정보 처리
    • 360° CDMO 및 CRO 서비스
    • CDMO 서비스
    • 임상시험 CRO 서비스
    • 장비 서비스
    • 교육 서비스
    • Unity Lab Services
    • 서비스 전체 보기
  • 지원
    • 고객센터
    • 문의하기
    • 시험성적서(COA) 및 적합성 인증서(COC)
    • Safety Data Sheets (SDS)
    • 매뉴얼
    • 인용 및 참고 문헌
    • 기기 지원
    • 기술 자료/제품 FAQ
    • 학습 센터
    • 지원 메뉴 전체보기
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 문의하기
  • 빠른 주문
  • 주문 현황
  • 제품 문서 검색
Thermo Fisher Scientific Logo

Search

전체검색
Search button
          • 주문 현황
          • 빠른주문
          • 로그인
            로그인
            회원이 아니신가요? 계정 생성하기
            • 계정정보
            • 주문내역조회
            • 커스텀제품 및 프로젝트
            • Services Central
          • Primary Antibodies ›
          • OCT4 Antibodies

          Invitrogen

          OCT4 Polyclonal Antibody

          Advanced Verification
          2 Published Figures
          4 References
          View all (67) OCT4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite OCT4 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          • Published Figures (2)
          • Advanced Verification (1)
          OCT4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          OCT4 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          OCT4 Antibody (PA1-16943) in ICC/IF

          Immunocytochemistry analysis of OCT4 in Ntera2 cells. Samples were incubated with OCT4 polyclonal antibody (Product # PA1-16943) using a dilution of 1 µg/mL overnight at 4°C followed by anti-rabbit Dylight 488 (Green) at a dilution of 1:1,000 dilution for 60 minutes. Cells were fixed in 4% paraformaldehyde for 10 minutes and permeabilized in 0.5% Triton X-100 in PBS for 5 minutes. Nuclei were counterstained with DAPI (Blue). Cells were imaged using a 100X objective and digitally deconvolved. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          OCT4 Antibody in Immunocytochemistry (ICC/IF)
          OCT4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OCT4 Antibody in Western Blot (WB)
          OCT4 Antibody in Western Blot (WB)
          OCT4 Antibody in Western Blot (WB)
          OCT4 Antibody in Flow Cytometry (Flow)
          OCT4 Antibody in Flow Cytometry (Flow)
          OCT4 Antibody in Flow Cytometry (Flow)
          OCT4 Antibody in Flow Cytometry (Flow)
          OCT4 Antibody in Immunocytochemistry (ICC/IF)
          OCT4 Antibody in Western Blot (WB)
          OCT4 Antibody
          OCT4 Polyclonal Antibody

          Product Details

          PA1-16943

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          1:50-1:200
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Non-human primate, Rat

          Published species

          Bovine, Cat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide made to an internal portion of the human OCT4 protein (between residues 100-200).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2268556

          Product Specific Information

          This antibody is expected to react with swine, feline and bovine samples based on a 93% sequence homology.

          Target Information

          POU5F1, also commonly known as Oct-4, is a maternally expressed octamer-binding protein that was the first transcription factor described for the early stages of development. The role of POU5F1 in embryonic development suggested that it might be useful in the creation of stem cells that might be useful in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing POU5F1 and the transcription factors Sox2, Klf4 and Lin28 along with c-Myc in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing POU5F1, Sox2, KlfF4 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 3-Oct; 4-Oct; DADB-104B20.2; gp-9; HGNC:9221; MGC22487; NF-A3; Oct-3; Oct-4; OCT4-B1; OCT4B-variant 2; octamer transcription factor 4; Octamer-binding protein 3; Octamer-binding protein 4; Octamer-binding transcription factor 3; OTF-3; OTTHUMP00000221150; OTTHUMP00000221151; POU class 5 homeobox 1; POU domain transcription factor OCT4; POU domain, class 5, transcription factor 1; POU transcription factor; Pou5f1; pou-2; POU-type homeodomain-containing DNA-binding protein; pou2; transcriptional regulator

          View more View less

          Gene Aliases: NF-A3; Oct-3; Oct-3/4; Oct-4; OCT3; Oct3/4; OCT4; OCT4Borf1; OTF-3; Otf-4; OTF3; Otf3-rs7; Otf3g; OTF4; POU5F1

          View more View less

          UniProt ID: (Human) Q01860, (Mouse) P20263

          View more View less

          Entrez Gene ID: (Bovine) 282316, (Human) 5460, (Mouse) 18999, (Rat) 294562

          View more View less

          Function(s)
          RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcription factor activity, sequence-specific DNA binding RNA binding protein binding ubiquitin protein ligase binding miRNA binding sequence-specific DNA binding RNA polymerase II sequence-specific DNA binding transcription factor binding sequence-specific double-stranded DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding RNA polymerase II transcription coactivator activity chromatin binding transcription corepressor activity transcription factor binding chromatin DNA binding transcription regulatory region DNA binding poly(A) RNA binding protein heterodimerization activity POU domain binding HMG box domain binding
          Process(es)
          regulation of transcription from RNA polymerase II promoter multicellular organism development negative regulation of transcription from RNA polymerase II promoter endodermal cell fate specification blastocyst development heart induction endodermal-mesodermal cell signaling regulation of transcription, DNA-templated response to wounding anatomical structure morphogenesis regulation of asymmetric cell division regulation of gene expression BMP signaling pathway somatic stem cell population maintenance positive regulation of transcription from RNA polymerase II promoter cell fate commitment involved in formation of primary germ layer cardiac cell fate determination positive regulation of canonical Wnt signaling pathway negative regulation of pri-miRNA transcription from RNA polymerase II promoter mesodermal cell fate commitment endodermal cell fate commitment ectodermal cell fate commitment trophectodermal cell differentiation blastocyst growth BMP signaling pathway involved in heart induction transcription, DNA-templated transcription from RNA polymerase II promoter response to organic substance stem cell population maintenance germ-line stem cell population maintenance response to retinoic acid positive regulation of catenin import into nucleus mRNA transcription from RNA polymerase II promoter cell fate commitment negative regulation of cell differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated negative regulation of calcium ion-dependent exocytosis stem cell differentiation positive regulation of protein kinase B signaling negative regulation of protein kinase B signaling positive regulation of SMAD protein import into nucleus negative regulation of gene silencing by miRNA regulation of heart induction by regulation of canonical Wnt signaling pathway regulation of methylation-dependent chromatin silencing response to cytokine
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          온라인 주문 Plus Icon Minus Icon
          • 주문 현황
          • 주문 지원
          • 빠른 주문
          • Supply Center
          • eProcurement
          지원 Plus Icon Minus Icon
          • Help and Support
          • 고객 센터
          • 기술 지원 센터
          • 제품 문서 검색
          • 사이트 문제 보고
          교육 및 이벤트 Plus Icon Minus Icon
          • 교육 센터
          • 프로모션
          • 이벤트 및 웨비나
          • 소셜 미디어
          About Thermo Fisher Plus Icon Minus Icon
          • 소개 소개
          • 채용 채용
          • 투자자 투자자
          • 뉴스 뉴스
          • 사회적 책임 사회적 책임
          • Trademarks
          • 공정거래 공정거래
          • 정책 및 고지
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • 이용 약관
          • 개인정보 처리방침
          • 가격 및 운임 정책
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat

          써모 피셔 사이언티픽 코리아 주식회사
          대표자 : 석수진
          사업자 등록번호 : 117-81-46910
          입금계좌 : 하나은행 336-890014-06204
          (예금주 : 써모피셔사이언티픽코리아 주식회사)

           

          써모 피셔 사이언티픽 솔루션스 유한회사
          대표자 : 석수진
          사업자 등록번호 : 114-86-04783
          입금계좌 : 신한은행 140-004-396660
          (예금주 : 써모피셔사이언티픽솔루션스 유한회사)

           

          주소 : 서울시 강남구 광평로 281 수서오피스빌딩 12층 06349 | 통신판매업신고번호 : 2015-서울강남-00898

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline