Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CCL22 may play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. Also, it is a chemotactic for monocytes. dendritic cells and natural killer cells. CCL22 is a mild chemoattractant for primary activated T-lymphocytes and a potent chemoattractant for chronically activated T-lymphocytes but has no chemattractant activity for neutrophils, eosinophils, and resting T-lymphocytes. Binds to CCR4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AW060611; CC chemokine STCP-1; chemokine (C-C motif) ligand 22; macrophage-derived chemokine; MDC(1-69); MGC108943; MGC34554; RP24-69A20.1; small inducible cytokine A22; small inducible cytokine subfamily A (Cys-Cys), member 22; stimulated T cell chemotactic protein 1; unnamed protein product
Gene Aliases: A-152E5.1; ABCD-1; DC/B-CK; MDC; SCYA22; STCP-1
UniProt ID: (Human) O00626
Entrez Gene ID: (Human) 6367
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support