Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SAP97 Antibodies

          Invitrogen

          SAP97 Polyclonal Antibody

          Advanced Verification
          26 Published Figures
          37 References
          View all (22) SAP97 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SAP97 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (8)
          • Published Figures (26)
          • Advanced Verification (6)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 40

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SAP97 Antibody (PA1-741) in ICC/IF

          Immunofluorescence analysis of SAP97 was performed using 70% confluent log phase RSC96 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with SAP97 Rabbit Polyclonal Antibody (Product # PA1-741) at 1:250 dilution in 0.1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Prod... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Immunocytochemistry (ICC/IF)
          SAP97 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          SAP97 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          SAP97 Antibody in Western Blot, Immunohistochemistry, Immunoprecipitation (WB, IHC, IP)
          SAP97 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          SAP97 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot (WB)
          SAP97 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          SAP97 Antibody in Immunoprecipitation (IP)
          SAP97 Antibody in Immunoprecipitation (IP)
          SAP97 Antibody
          SAP97 Antibody
          SAP97 Antibody
          SAP97 Antibody
          SAP97 Antibody
          SAP97 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SAP97 Polyclonal Antibody

          Product Details

          PA1-741

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:5,000
          View 19 publications 19 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Frozen) (IHC (F))

          1:1,000
          -

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:200 - 1:250
          View 6 publications 6 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 5 publications 5 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues V(115) L P S E R I S P Q V P N E V L G P E(133) of rat SAP97.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Purification

          Ammonium sulfate precipitation

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2092020

          Product Specific Information

          PA1-741 detects Synapse-Associated Protein 97 (SAP97) from human, rat and mouse tissues. No cross-reactivity to other proteins, including synapse-associated protein family members, has been observed.

          PA1-741 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, immunoprecipitation, and immunohistochemistry procedures. By Western blot, this antibody detects an ~140 kDa protein representing SAP97 from rat brain extract. Immunohistochemical staining of SAP97 in rat hippocampus with PA1-741 results in intense neuronal cell body and dendrite staining.

          The PA1-741 immunizing peptide corresponds to amino acids 115-133 from rat SAP97. This sequence is 84% and 74% conserved in mouse and human SAP97, respectively.

          PA1-741 can be used with blocking peptide PEP-055.

          Target Information

          This gene encodes a multi-domain scaffolding protein that is required for normal development. This protein may have a role in septate junction formation, signal transduction, cell proliferation, synaptogenesis and lymphocyte activation. Several alternatively spliced transcript variants encoding different isoforms have been described for this gene, but the full-length nature of some of the variants is not known.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: discs large homolog 1, scribble cell polarity complex component; discs, large homolog 1; dJ1061C18.1.1; DKFZp761P0818; DKFZp781B0426; DLG1; E-dlg/SAP97; Drosophila discs-large tumor suppressor homologue (synapse associated protein); embryo-dlg/synapse-associated protein 97; presynaptic protein SAP97; synapse-associated protein 97; unnamed protein product

          View more View less

          Gene Aliases: B130052P05Rik; DLGH1; E-dlg/SAP97; hdlg; mKIAA4187; SAP-97; SAP97

          View more View less

          UniProt ID: (Human) Q12959, (Rat) Q62696, (Mouse) Q811D0

          View more View less

          Entrez Gene ID: (Human) 1739, (Rat) 25252, (Mouse) 13383

          View more View less

          Function(s)
          guanylate kinase activity phosphoprotein phosphatase activity protein binding cytoskeletal protein binding potassium channel regulator activity kinase binding protein kinase binding phosphatase binding ionotropic glutamate receptor binding ion channel binding cadherin binding binding, bridging L27 domain binding structural constituent of postsynaptic density kinesin binding enzyme binding PDZ domain binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter endothelial cell proliferation actin filament organization establishment or maintenance of cell polarity chemical synaptic transmission nervous system development regulation of cell shape cortical actin cytoskeleton organization astral microtubule organization protein complex localization regulation of protein localization protein localization to synapse regulation of membrane potential receptor clustering regulation of potassium ion transport positive regulation of potassium ion transport cortical microtubule organization establishment or maintenance of epithelial cell apical/basal polarity GMP metabolic process GDP metabolic process establishment of centrosome localization negative regulation of ERK1 and ERK2 cascade bicellular tight junction assembly protein localization to membrane protein localization to plasma membrane receptor localization to synapse cell-cell adhesion regulation of ventricular cardiac muscle cell action potential membrane repolarization during ventricular cardiac muscle cell action potential regulation of postsynaptic membrane neurotransmitter receptor levels maintenance of postsynaptic density structure neurotransmitter receptor localization to postsynaptic specialization membrane regulation of NIK/NF-kappaB signaling regulation of sodium ion transmembrane transport regulation of protein localization to synapse positive regulation of protein localization to plasma membrane regulation of potassium ion import negative regulation of p38MAPK cascade regulation of potassium ion export across plasma membrane protein localization to cell periphery negative regulation of G1/S transition of mitotic cell cycle ureteric bud development branching involved in ureteric bud morphogenesis immunological synapse formation lens development in camera-type eye cell adhesion protein localization positive regulation of cell proliferation positive regulation of gene expression epithelial structure maintenance cerebral cortex development peristalsis positive regulation of actin filament polymerization membrane raft organization regulation of myelination activation of protein kinase activity T cell activation negative regulation of T cell proliferation amyloid precursor protein metabolic process reproductive structure development embryonic skeletal system morphogenesis tissue morphogenesis smooth muscle tissue development negative regulation of epithelial cell proliferation negative regulation of protein kinase B signaling hard palate development cellular response to brain-derived neurotrophic factor stimulus positive regulation of multicellular organism growth positive regulation of developmental growth
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch